Learn More
Biomatik Corporation Mouse Tspo (Translocator protein) ELISA Kit
Supplier:  Biomatik Corporation EKF59060
Encompass_Preferred
Mouse Tspo (Translocator protein) ELISA Kit for detecting Tspo in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P50637. Target Synonyms: Tspo; Translocator protein; PBR; Peripheral-type benzodiazepine receptor; Mitochondrial benzodiazepine receptor; PKBS; Bzrp; Mbr. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8235