Learn More
Biomatik Corporation Human CHEM (Chemerin) ELISA Kit
Supplier:  Biomatik Corporation EKF59126
Encompass_Preferred
Human CHEM (Chemerin) ELISA Kit for detecting CHEM in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q99969. Target Synonyms: RARRES2; TIG-2; chemerin; CHEM; RARRES2; RAR-responsive protein TIG2; retinoic acid receptor responder; tazarotene induced 2; retinoic acid receptor responder protein 2; Tazarotene-induced gene 2 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8242