Learn More
Biomatik Corporation Bovine IPTH (Intact PaRathormone) ELISA Kit
Supplier:  Biomatik Corporation EKF59235
Encompass_Preferred
Bovine IPTH (Intact PaRathormone) ELISA Kit for detecting IPTH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01268. Target Synonyms: IPTH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8256