Learn More
Biomatik Corporation Human Selenoprotein P1, Plasma (SEPP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07255
Encompass_Preferred
Human Selenoprotein P1, Plasma (SEPP1) ELISA Kit for detecting SEPP1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49908. Target Synonyms: SEPP; SeP; SEP-P1; SELP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3771