Learn More
Biomatik Corporation Human Semaphorin 3F (SEMA3F) ELISA Kit, 96 tests
Catalog No. 501503778
Encompass_Preferred
Human Semaphorin 3F (SEMA3F) ELISA Kit for detecting SEMA3F in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13275. Gene ID: 6405. Target Synonyms: SEMA-IV; SEMA4; SEMAK; Sema Domain,Immunoglobulin Domain (Ig) ,Transmembrane Domain (TM) and Short Cytoplasmic Domain 3F. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3778 Supplier Biomatik Corporation Supplier No. EKU07264
Human Semaphorin 3F (SEMA3F) ELISA Kit for detecting SEMA3F in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13275. Gene ID: 6405. Target Synonyms: SEMA-IV; SEMA4; SEMAK; Sema Domain,Immunoglobulin Domain (Ig) ,Transmembrane Domain (TM) and Short Cytoplasmic Domain 3F. Restrictions: For Research Use Only. Not for use in diagnostic procedures.