Learn More
Biomatik Corporation Human Semaphorin 4D (SEMA4D) ELISA Kit, 96 tests
Catalog No. 501503780
Encompass_Preferred
Human Semaphorin 4D (SEMA4D) ELISA Kit for detecting SEMA4D in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92854. Gene ID: 10507. Target Synonyms: CD100; A8; BB18; GR3; M-Sema G; M-Sema-G; SEMAJ; Coll-4; Sema Domain,Immunoglobulin Domain (Ig) ,Transmembrane Domain (TM) and Short Cytoplasmic Domain 4D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3780 Supplier Biomatik Corporation Supplier No. EKU07267
Human Semaphorin 4D (SEMA4D) ELISA Kit for detecting SEMA4D in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92854. Gene ID: 10507. Target Synonyms: CD100; A8; BB18; GR3; M-Sema G; M-Sema-G; SEMAJ; Coll-4; Sema Domain,Immunoglobulin Domain (Ig) ,Transmembrane Domain (TM) and Short Cytoplasmic Domain 4D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.