Learn More
Biomatik Corporation Rat CB1R/CNR1 (Cannabinoid receptor 1) ELISA Kit
Supplier:  Biomatik Corporation EKF59767
Encompass_Preferred
Rat CB1R/CNR1 (Cannabinoid receptor 1) ELISA Kit for detecting CB1R; CNR1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P20272. Target Synonyms: Cannabinoid receptor 1; CB-R; CB1; Brain-type cannabinoid receptor; Skr6; Cnr1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8284