Learn More
Biomatik Corporation Human Semaphorin 5A (SEMA5A) ELISA Kit, 96 tests
Catalog No. 501503781
Encompass_Preferred
Human Semaphorin 5A (SEMA5A) ELISA Kit for detecting SEMA5A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.33ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13591. Gene ID: 9037. Target Synonyms: SEMAF; SemF; Semaphorin-F; Sema Domain,Immunoglobulin Domain (Ig) ,Transmembrane Domain (TM) and Short Cytoplasmic Domain 5A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3781 Supplier Biomatik Corporation Supplier No. EKU07268
Human Semaphorin 5A (SEMA5A) ELISA Kit for detecting SEMA5A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.33ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13591. Gene ID: 9037. Target Synonyms: SEMAF; SemF; Semaphorin-F; Sema Domain,Immunoglobulin Domain (Ig) ,Transmembrane Domain (TM) and Short Cytoplasmic Domain 5A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.