Learn More
Biomatik Corporation Rat Pvalb (Parvalbumin alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF60362
Encompass_Preferred
Rat Pvalb (Parvalbumin alpha) ELISA Kit for detecting Pvalb in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02625. Target Synonyms: Pvalb; PVALB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8300