Learn More
Biomatik Corporation Human ALDH4A1 (Delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial) ELISA Kit
Supplier:  Biomatik Corporation EKF60364
Encompass_Preferred
Human ALDH4A1 (Delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial) ELISA Kit for detecting ALDH4A1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P30038. Target Synonyms: ALDH4A1; Delta-1-pyrroline-5-carboxylate dehydrogenase; mitochondrial; P5C dehydrogenase; Aldehyde dehydrogenase family 4 member A1; L-glutamate gamma-semialdehyde dehydrogenase; ALDH4; P5CDH; P5CD; P5CDh; P5CDhL; P5CDhS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8302