Learn More
Biomatik Corporation Human Serine Peptidase Inhibitor Kazal Type 1 (SPINK1) ELISA Kit, 96 tests
Catalog No. 501503787
Encompass_Preferred
Human Serine Peptidase Inhibitor Kazal Type 1 (SPINK1) ELISA Kit for detecting SPINK1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00995. Gene ID: 6690. Target Synonyms: PCTT; PSTI; TATI; Tumor-Associated Trypsin Inhibitor; Pancreatic secretory trypsin inhibitor; Tumor-associated trypsin inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3787 Supplier Biomatik Corporation Supplier No. EKU07285
Human Serine Peptidase Inhibitor Kazal Type 1 (SPINK1) ELISA Kit for detecting SPINK1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00995. Gene ID: 6690. Target Synonyms: PCTT; PSTI; TATI; Tumor-Associated Trypsin Inhibitor; Pancreatic secretory trypsin inhibitor; Tumor-associated trypsin inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.