Learn More
Biomatik Corporation Human GH2 (Growth Hormone 2) ELISA Kit
Supplier:  Biomatik Corporation EKF60370
Encompass_Preferred
Human GH2 (Growth Hormone 2) ELISA Kit for detecting GH2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01242. Target Synonyms: GH2; GHL; GHV; GHL; GHVgrowth hormone variant; GH-VPlacenta-specific growth hormone; growth hormone 2hGH-V; placental-specific growth hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8307