Learn More
Biomatik Corporation Human IGFBP-3 (Insulin-like growth factor-binding protein 3) ELISA Kit
Supplier:  Biomatik Corporation EKF60374
Encompass_Preferred
Human IGFBP-3 (Insulin-like growth factor-binding protein 3) ELISA Kit for detecting IGFBP-3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.391-25ng/ml. Sensitivity: 0.234ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P17936. Target Synonyms: IGFBP-3; IGFBP3; BP-53; IBP3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8311