Learn More
Biomatik Corporation Human Serine Peptidase Inhibitor Kunitz Type 2 (SPINT2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07288
Encompass_Preferred
Human Serine Peptidase Inhibitor Kunitz Type 2 (SPINT2) ELISA Kit for detecting SPINT2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.134ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O43291. Gene ID: 10653. Target Synonyms: HAI2; Kop; PB; Placental Bikunin; Hepatocyte growth factor activator inhibitor type 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3789