Learn More
Biomatik Corporation Human Serotonin Transporter (SERT) ELISA Kit, 96 tests
Catalog No. 501503790
Encompass_Preferred
Human Serotonin Transporter (SERT) ELISA Kit for detecting SERT in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31645. Gene ID: 6532. Target Synonyms: SLC6A4; SLC6-A4; HTT; 5-HTT; 5HTT; OCD1; hSERT; Solute Carrier Family 6,Member 4; Neurotransmitter Transporter,Serotonin; Sodium-dependent serotonin transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3790 Supplier Biomatik Corporation Supplier No. EKU07293
Add to Cart
Add to Cart
Human Serotonin Transporter (SERT) ELISA Kit for detecting SERT in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31645. Gene ID: 6532. Target Synonyms: SLC6A4; SLC6-A4; HTT; 5-HTT; 5HTT; OCD1; hSERT; Solute Carrier Family 6,Member 4; Neurotransmitter Transporter,Serotonin; Sodium-dependent serotonin transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.