Learn More
Biomatik Corporation Human AKR1B10 (Aldo-keto reductase family 1 member B10) ELISA Kit
Supplier:  Biomatik Corporation EKF60377
Encompass_Preferred
Human AKR1B10 (Aldo-keto reductase family 1 member B10) ELISA Kit for detecting AKR1B10 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O60218. Target Synonyms: AKR1B10; Aldo-keto reductase family 1 member B10; Small intestine reductase; SI reductase; Aldose reductase-related protein; ARP; hARP; ARL-1; Aldose reductase-like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8314