Learn More
Biomatik Corporation Cattle Serum Amyloid A (SAA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07296
Encompass_Preferred
The Cattle Serum Amyloid A (SAA) ELISA Kit allows for quantitative determination of Serum Amyloid A in Serum, Plasma and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.067 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P35541. Cross Activity: No significant cross-reactivity or interference between Serum Amyloid A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3792