Learn More
Biomatik Corporation Rat Cpb1 (Carboxypeptidase B) ELISA Kit
Supplier:  Biomatik Corporation EKF60379
Encompass_Preferred
Rat Cpb1 (Carboxypeptidase B) ELISA Kit for detecting Cpb1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P19223. Target Synonyms: Cpb1; PASP; PCPB; carboxypeptidase B1; tissue; CPB; DKFZp779K1333; EC 3.4.17; EC 3.4.17.2; Pancreas-specific protein; PASP; PCPB; procarboxypeptidase B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8316