Learn More
Biomatik Corporation Human GAL9 (Galectin-9) ELISA Kit
Supplier:  Biomatik Corporation EKF60380
Encompass_Preferred
Human GAL9 (Galectin-9) ELISA Kit for detecting GAL9 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O00182. Target Synonyms: Galectin-9; GAL9; Galectin 9; Ecalectin; LGALS9; ecalectin; Ecalectin; Gal-9; gal-9; HUAT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8317