Learn More
Biomatik Corporation Mouse Sestrin 1 (SESN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07306
Encompass_Preferred
Mouse Sestrin 1 (SESN1) ELISA Kit for detecting SESN1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P58006. Gene ID: 140742. Target Synonyms: SEST1; PA26; p53-regulated protein PA26. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3795