Learn More
Biomatik Corporation Human Sestrin 2 (SESN2) ELISA Kit, 96 tests
Catalog No. 501503796
Encompass_Preferred
Human Sestrin 2 (SESN2) ELISA Kit for detecting SESN2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P58004. Gene ID: 83667. Target Synonyms: HI95; SES2; SEST2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3796 Supplier Biomatik Corporation Supplier No. EKU07307
Human Sestrin 2 (SESN2) ELISA Kit for detecting SESN2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P58004. Gene ID: 83667. Target Synonyms: HI95; SES2; SEST2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.