Learn More
Biomatik Corporation Human ANXA3 (Annexin A3) ELISA Kit
Supplier:  Biomatik Corporation EKF60389
Encompass_Preferred
Human ANXA3 (Annexin A3) ELISA Kit for detecting ANXA3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P12429. Target Synonyms: ANXA3; Annexin A3; Calcimedin 35-alpha; Lipocortin III; PAP-III; 35-alpha calcimedin; annexin A3; 1; 2-cyclic-inositol-phosphate phosphodiesterase; Annexin III; Annexin-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8325