Learn More
Biomatik Corporation Mouse Sestrin 3 (SESN3) ELISA Kit, 96 tests
Catalog No. 501503797
Encompass_Preferred
Mouse Sestrin 3 (SESN3) ELISA Kit for detecting SESN3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9CYP7. Gene ID: 75747. Target Synonyms: SEST3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3797 Supplier Biomatik Corporation Supplier No. EKU07309
Mouse Sestrin 3 (SESN3) ELISA Kit for detecting SESN3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9CYP7. Gene ID: 75747. Target Synonyms: SEST3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.