Learn More
Biomatik Corporation Human IL20RA (Interleukin-20 receptor subunit alpha) ELISA Kit
Supplier:  Biomatik Corporation EKF60396
Encompass_Preferred
Human IL20RA (Interleukin-20 receptor subunit alpha) ELISA Kit for detecting IL20RA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9UHF4. Target Synonyms: IL20RA; CRF2-8; zcytor7; class II cytokine receptor ZCYTOR7; Cytokine receptor class-II member 8; Cytokine receptor family 2 member 8; IL-20R1; IL-20 receptor subunit alpha; IL-20RA; IL-20R-alpha; interleukin 20 receptor; alpha; interleukin-20 receptor I; interleukin-20 receptor subunit alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8330