Learn More
Biomatik Corporation Human Sex Hormone Binding Globulin (SHBG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07316
Encompass_Preferred
Human Sex Hormone Binding Globulin (SHBG) ELISA Kit for detecting SHBG in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04278. Gene ID: 6462. Target Synonyms: ABP; TEBG; SBP; Sex steroid-binding protein; Testis-specific androgen-binding protein; Testosterone-estradiol-binding globulin; Testosterone-estrogen-binding globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3799