Learn More
Biomatik Corporation Human RNASET2 (Ribonuclease T2) ELISA Kit
Supplier:  Biomatik Corporation EKF60401
Encompass_Preferred
Human RNASET2 (Ribonuclease T2) ELISA Kit for detecting RNASET2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O00584. Target Synonyms: RNASET2; RNASE6PL; Ribonuclease T2; Ribonuclease 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8335