Learn More
Biomatik Corporation Mouse Sialic Acid Binding Ig Like Lectin 3 (SIGLEC3) ELISA Kit, 96 tests
Catalog No. 501503801
Encompass_Preferred
Mouse Cluster Of Differentiation 33 (CD33) ELISA Kit for detecting CD33 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63994. Gene ID: 12489. Target Synonyms: SIGLEC3; Gp67; p67; Myeloid cell surface antigen CD33. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3801 Supplier Biomatik Corporation Supplier No. EKU07328
Mouse Cluster Of Differentiation 33 (CD33) ELISA Kit for detecting CD33 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63994. Gene ID: 12489. Target Synonyms: SIGLEC3; Gp67; p67; Myeloid cell surface antigen CD33. Restrictions: For Research Use Only. Not for use in diagnostic procedures.