Learn More
Biomatik Corporation Human Signaling Lymphocytic Activation Molecule Family, Member 7 (SLAMF7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07346
Encompass_Preferred
Human Signaling Lymphocytic Activation Molecule Family, Member 7 (SLAMF7) ELISA Kit for detecting SLAMF7 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NQ25. Gene ID: 57823. Target Synonyms: CD319; CRACC; 19A; CS1; CD2-like receptor-activating cytotoxic cells; Membrane protein FOAP-12. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3803