Learn More
Biomatik Corporation Human G-CSF (Granulocyte Colony Stimulating Factor 3) ELISA Kit
Supplier:  Biomatik Corporation EKF60438
Encompass_Preferred
Human G-CSF (Granulocyte Colony Stimulating Factor 3) ELISA Kit for detecting G-CSF in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 39.063-2500pg/ml. Sensitivity: 23.438pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P09919. Target Synonyms: G-CSF; CSF3; C17orf33; CSF3OS; Pluripoietin; Filgrastim; Lenograstim. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8350