Learn More
Biomatik Corporation Mouse F11 (Coagulation factor XI) ELISA Kit
Supplier:  Biomatik Corporation EKF60443
Encompass_Preferred
Mouse F11 (Coagulation factor XI) ELISA Kit for detecting F11 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q91Y47. Target Synonyms: F11; Coagulation factor XI; FXI; Plasma thromboplastin antecedent; PTA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8352