Learn More
Biomatik Corporation Human SLC34A2 (Sodium-dependent phosphate transport protein 2B) ELISA Kit
Supplier:  Biomatik Corporation EKF60463
Encompass_Preferred
Human SLC34A2 (Sodium-dependent phosphate transport protein 2B) ELISA Kit for detecting SLC34A2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O95436. Target Synonyms: Sodium-dependent phosphate transport protein 2B; Sodium-phosphate transport protein 2B; Na(+)-dependent phosphate cotransporter 2B; NaPi3b; Sodium/phosphate cotransporter 2B; Na(+)/Pi cotransporter 2B; NaPi-2b; Solute carrier family 34 member 2; SLC34A2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8363