Learn More
Biomatik Corporation Human IL-22 (Interleukin-22) ELISA Kit
Supplier:  Biomatik Corporation EKF60479
Encompass_Preferred
Human IL-22 (Interleukin-22) ELISA Kit for detecting IL-22 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9GZX6. Target Synonyms: IL-22; IL22; IL-D110; IL-TIF; ILTIF; TIFIL-23; TIFa; zcyto18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8367