Learn More
Biomatik Corporation Mouse Sjogren Syndrome Antigen B (SSB) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07361
Encompass_Preferred
Mouse Sjogren Syndrome Antigen B (SSB) ELISA Kit for detecting SSB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32067. Gene ID: 20823. Target Synonyms: LARP3; Lupus La Protein; Autoantigen La; La Ribonucleoprotein Domain Family,Member 3; Autoantigen SSB/La. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3811