Learn More
Biomatik Corporation Human Slit Homolog 2 (Slit2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07368
Encompass_Preferred
Human Slit Homolog 2 (Slit2) ELISA Kit for detecting Slit2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O94813. Gene ID: 9353. Target Synonyms: SLIL3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3813