Learn More
Biomatik Corporation Human Solute Carrier Family 16, Member 1 (SLC16A1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07391
Encompass_Preferred
Human Solute Carrier Family 16, Member 1 (SLC16A1) ELISA Kit for detecting SLC16A1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P53985. Gene ID: 6566. Target Synonyms: MCT1; MCT; Monocarboxylate Transporter 1; Monocarboxylic Acid Transporter 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3815