Learn More
Biomatik Corporation Rat D2D (D-Dimer) ELISA Kit
Supplier:  Biomatik Corporation EKF60662
Encompass_Preferred
Rat D2D (D-Dimer) ELISA Kit for detecting D2D in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.813-500ng/ml. Sensitivity: 4.688ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P06399; P14480; P14480. Target Synonyms: D-Dimer; D2D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8381