Learn More
Biomatik Corporation Rat Solute Carrier Family 30, Member 1 (SLC30A1) ELISA Kit, 96 tests
Catalog No. 501503816
Encompass_Preferred
Rat Solute Carrier Family 30, Member 1 (SLC30A1) ELISA Kit for detecting SLC30A1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62720. Gene ID: 58976. Target Synonyms: ZNT1; ZRC1; Zinc Transporter 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3816 Supplier Biomatik Corporation Supplier No. EKU07401
Rat Solute Carrier Family 30, Member 1 (SLC30A1) ELISA Kit for detecting SLC30A1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62720. Gene ID: 58976. Target Synonyms: ZNT1; ZRC1; Zinc Transporter 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.