Learn More
Biomatik Corporation Chicken IL-10 (Interleukin-10) ELISA Kit
Supplier:  Biomatik Corporation EKF60699
Encompass_Preferred
Chicken IL-10 (Interleukin-10) ELISA Kit for detecting IL-10 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q6A2H4. Target Synonyms: IL-10; IL10; IL10A; CSIF; TGIF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8388