Learn More
Biomatik Corporation Human Somatostatin (SST) ELISA Kit, 96 tests
Catalog No. 501503818
Encompass_Preferred
Human Somatostatin (SST) ELISA Kit for detecting SST in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.17-500pg/ml. Sensitivity: 2.62pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P61278. Gene ID: 6750. Target Synonyms: GHIH; SRIF; SMST; GHRIH; GHRIF; Growth Hormone Releasing Inhibiting Hormone; Somatotropin Release-Inhibiting Factor; Somatostatin-14; Somatostatin-28. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3818 Supplier Biomatik Corporation Supplier No. EKU07410
Human Somatostatin (SST) ELISA Kit for detecting SST in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.17-500pg/ml. Sensitivity: 2.62pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P61278. Gene ID: 6750. Target Synonyms: GHIH; SRIF; SMST; GHRIH; GHRIF; Growth Hormone Releasing Inhibiting Hormone; Somatotropin Release-Inhibiting Factor; Somatostatin-14; Somatostatin-28. Restrictions: For Research Use Only. Not for use in diagnostic procedures.