Learn More
Biomatik Corporation Mouse Somatostatin (SST) ELISA Kit, 96 tests
Catalog No. 501503819
Encompass_Preferred
Mouse Somatostatin (SST) ELISA Kit for detecting SST in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.77pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P60041. Gene ID: 20604. Target Synonyms: GHIH; SRIF; SMST; GHRIH; GHRIF; Growth Hormone Releasing Inhibiting Hormone; Somatotropin Release-Inhibiting Factor; Somatostatin-14; Somatostatin-28. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3819 Supplier Biomatik Corporation Supplier No. EKU07411
Mouse Somatostatin (SST) ELISA Kit for detecting SST in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.77pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P60041. Gene ID: 20604. Target Synonyms: GHIH; SRIF; SMST; GHRIH; GHRIF; Growth Hormone Releasing Inhibiting Hormone; Somatotropin Release-Inhibiting Factor; Somatostatin-14; Somatostatin-28. Restrictions: For Research Use Only. Not for use in diagnostic procedures.