Learn More
Biomatik Corporation Monkey C5a (Complement Component 5a) ELISA Kit
Supplier:  Biomatik Corporation EKF60705
Encompass_Preferred
Monkey C5a (Complement Component 5a) ELISA Kit for detecting C5a in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: A0A2K5U1T1. Target Synonyms: C5a. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8393