Learn More
Biomatik Corporation Monkey IL-2 (Interleukin 2) ELISA Kit
Supplier:  Biomatik Corporation EKF60710
Encompass_Preferred
Monkey IL-2 (Interleukin 2) ELISA Kit for detecting IL-2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q7JFM5. Target Synonyms: IL-2; Interleukin 2; IL2; TCGF; Lymphokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8398