Learn More
Biomatik Corporation Human Spleen Tyrosine Kinase (SYK) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07442
Encompass_Preferred
Human Spleen Tyrosine Kinase (SYK) ELISA Kit for detecting SYK in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43405. Gene ID: 6850. Target Synonyms: p72-Syk; Spleen tyrosine kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3826