Learn More
Biomatik Corporation Mouse Splicing Factor 3B Subunit 3 (SF3B3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07443
Encompass_Preferred
Mouse Splicing Factor 3B Subunit 3 (SF3B3) ELISA Kit for detecting SF3B3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q921M3. Gene ID: 101943. Target Synonyms: RSE1; SAP130; SF3b130; STAF130; Spliceosome-associated protein 130; Pre-mRNA-splicing factor SF3b 130 kDa subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3827