Learn More
Biomatik Corporation Rat Spondin 2 (SPON2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07447
Encompass_Preferred
Rat Spondin 2 (SPON2) ELISA Kit for detecting SPON2 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WV75. Gene ID: 171569. Target Synonyms: DIL1; Mindin; M-spondin; Extracellular Matrix Protein; Differentially expressed in cancerous and non-cancerous lung cells 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3829