Learn More
Biomatik Corporation Human Squalene Epoxidase (SQLE) ELISA Kit, 96 tests
Catalog No. 501503830
Encompass_Preferred
Human Squalene Epoxidase (SQLE) ELISA Kit for detecting SQLE in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14534. Gene ID: 6713. Target Synonyms: SE; ERG1; Squalene monooxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3830 Supplier Biomatik Corporation Supplier No. EKU07448
Human Squalene Epoxidase (SQLE) ELISA Kit for detecting SQLE in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14534. Gene ID: 6713. Target Synonyms: SE; ERG1; Squalene monooxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.