Learn More
Biomatik Corporation Human Squamous Cell Carcinoma Antigen 2 (SCCA2) ELISA Kit, 96 tests
Catalog No. 501503832
Encompass_Preferred
Human Serpin Peptidase Inhibitor,Clade B,Member 4 (SERPINB4) ELISA Kit for detecting SERPINB4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86W05. Target Synonyms: SCCA2; PI11; SPB4; Leupin; Serpin Peptidase Inhibitor,Clade B,Member 4; Peptidase Inhibitor 11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3832 Supplier Biomatik Corporation Supplier No. EKU07450
Human Serpin Peptidase Inhibitor,Clade B,Member 4 (SERPINB4) ELISA Kit for detecting SERPINB4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q86W05. Target Synonyms: SCCA2; PI11; SPB4; Leupin; Serpin Peptidase Inhibitor,Clade B,Member 4; Peptidase Inhibitor 11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.