Learn More
Biomatik Corporation Human HNRNPA2B1 (Heterogeneous nuclear ribonucleoproteins A2/B1) ELISA Kit
Supplier:  Biomatik Corporation EKF60759
Encompass_Preferred
Human HNRNPA2B1 (Heterogeneous nuclear ribonucleoproteins A2/B1) ELISA Kit for detecting HNRNPA2B1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P22626. Target Synonyms: HNRPA2B1; Heterogeneous Nuclear Ribonucleoprotein A2; B1; HNRNPA2B1; hnRNP A2B1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8431