Learn More
Biomatik Corporation Human Stanniocalcin 1 (STC1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07452
Encompass_Preferred
Human Stanniocalcin 1 (STC1) ELISA Kit for detecting STC1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P52823. Gene ID: 6781. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3833