Learn More
Biomatik Corporation Human Statherin (STATH) ELISA Kit, 96 tests
Catalog No. 501503834
Encompass_Preferred
Human Statherin (STATH) ELISA Kit for detecting STATH in tissue homogenates, saliva, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02808. Gene ID: 6779. Target Synonyms: STR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3834 Supplier Biomatik Corporation Supplier No. EKU07456
Human Statherin (STATH) ELISA Kit for detecting STATH in tissue homogenates, saliva, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02808. Gene ID: 6779. Target Synonyms: STR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.